Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199098.1 | NP_954529.1 | 6 | 1208 | ||
XM_006241347.2 | XP_006241409.1 | 7 | 1305 | |||
XM_006241348.2 | XP_006241410.1 | 7 | 1158 | |||
XM_006241349.1 | XP_006241411.1 | 7 | 1043 | |||
GenBank mRNA | BC061871.1 | - | 6 | 1208 | 5598899 |