Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012133.5 | NP_036265.3 | 22 | 2423 | ||
GenBank mRNA | AB047847.1 | - | 22 | 2333 | ||
AF157833.1 | - | 22 | 2393 | |||
AF207598.1 | - | 22 | 2389 | |||
AF308295.1 | - | 3 | 286 | |||
AK002086.1 | - | 14 | 1725 | |||
AK022934.1 | - | 22 | 2320 | |||
AK291409.1 | - | 22 | 2413 | |||
BC017443.1 | - | 22 | 2316 |