Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Laptm4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199384.2 | NP_955416.1 | 1, 2 | 187 | ||
GenBank mRNA | BC063179.1 | - | 1, 2 | 191 | 6890564 | |
FQ212732.1 | - | 1, 2 | 589 | |||
FQ213363.1 | - | 1, 2 | 221 | |||
FQ232171.1 | - | 1, 2 | 245 | |||
FQ232930.1 | - | 1, 2 | 205 | |||
FQ235113.1 | - | 1, 2 | 92 |