Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flcn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199390.2 | NP_955422.1 | 12 | 1868 | ||
XM_006246489.2 | XP_006246551.1 | 12 | 1758 | |||
XM_006246490.2 | XP_006246552.1 | 12 | 1885 | |||
XM_006246491.1 | XP_006246553.1 | 13 | 2649 | |||
XM_008767816.1 | XP_008766038.1 | 12 | 1761 | |||
GenBank mRNA | AB096213.1 | - | 12 | 1751 | ||
BC085848.1 | - | 12 | 1868 | 7189414 | ||
FQ212214.1 | - | 12 | 1884 |