Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgef25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199395.1 | NP_955427.1 | 2, 3 | 242 | ||
XM_006241454.2 | XP_006241516.1 | 2, 3 | 554 | |||
XM_006241455.2 | XP_006241517.1 | 2, 3 | 556 | |||
XM_006241458.2 | XP_006241520.1 | 2, 3 | 277 | |||
GenBank mRNA | BC061879.1 | - | 2, 3 | 242 | 5599277 |