Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GALK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001556.2 | NP_001001556.1 | 2 | 337 | ||
NM_001289030.1 | NP_001275959.1 | 3 | 569 | |||
NM_001289031.1 | NP_001275960.1 | 2 | 422 | |||
NM_002044.3 | NP_002035.1 | 2 | 439 | |||
XM_005254279.3 | XP_005254336.1 | 2 | 228 | |||
XM_005254280.3 | XP_005254337.1 | 2 | 319 | |||
XM_005254284.3 | XP_005254341.1 | 2 | 319 | |||
XM_006720461.3 | XP_006720524.1 | 2 | 228 | |||
XM_006720462.3 | XP_006720525.1 | 2 | 209 | |||
XM_006720463.3 | XP_006720526.1 | 2 | 208 | |||
XM_006720464.3 | XP_006720527.1 | 2 | 208 | |||
XM_011521441.2 | XP_011519743.1 | 2 | 287 | |||
XM_017022062.1 | XP_016877551.1 | 2 | 209 | |||
XM_017022063.1 | XP_016877552.1 | 2 | 319 | |||
XM_017022065.1 | XP_016877554.1 | 2 | 339 | |||
XM_017022066.1 | XP_016877555.1 | 2 | 360 | |||
GenBank mRNA | AF461816.1 | - | 2 | 317 | ||
AK098049.1 | - | 2 | 178 | |||
AK301361.1 | - | 2 | 192 | |||
AK316440.1 | - | 3 | 303 | |||
AL832745.1 | - | 2 | 316 | |||
BC005141.2 | - | 2 | 207 | 3504930 | ||
BI766538.1 | - | 2 | 229 | |||
BQ921903.1 | - | 2 | 150 | |||
BT006901.1 | - | 2 | 132 | |||
DB457948.1 | - | 2 | 439 | |||
DC298656.1 | - | 2 | 429 | |||
DC390737.1 | - | 3 | 538 | |||
M84443.1 | - | 2 | 152 |