Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERGEF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012139.3 | NP_036271.1 | 3 | 362 | ||
NR_104040.1 | 3 | 362 | ||||
NR_104041.1 | 3 | 362 | ||||
GenBank mRNA | AJ243950.1 | - | 3 | 228 | ||
AJ243951.1 | - | 3 | 228 | |||
AK292286.1 | - | 3 | 362 | |||
AK294022.1 | - | 3 | 227 | |||
BC000707.2 | - | 3 | 236 | |||
BC065375.1 | - | 3 | 342 | 6672338 | ||
BU932034.1 | - | 3 | 340 |