Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199402.1 | NP_955434.1 | 2 | 215 | ||
XM_006247200.2 | XP_006247262.1 | 2 | 241 | |||
GenBank mRNA | AY438568.1 | - | 2 | 215 | ||
BC128718.1 | - | 2 | 175 | |||
BC158571.1 | - | 2 | 174 |