Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aqp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142366.2 | NP_001135838.1 | 5 | 1280 | ||
NM_001270558.2 | NP_001257487.1 | 4 | 1107 | |||
NM_001270559.2 | NP_001257488.1 | 4 | 1115 | |||
NM_012825.4 | NP_036957.1 | 5 | 1272 | |||
GenBank mRNA | DV717820.1 | - | 1 | 174 | ||
U14007.1 | - | 5 | 1206 |