Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171713.1 | NP_001165184.1 | 4, 5 | 271 | ||
NM_012157.3 | NP_036289.3 | 4, 5 | 274 | |||
XM_005265014.2 | XP_005265071.1 | 4, 5 | 294 | |||
XM_005265015.3 | XP_005265072.1 | 4, 5 | 495 | |||
XM_005265016.2 | XP_005265073.1 | 4, 5 | 239 | |||
XM_005265017.3 | XP_005265074.1 | 4, 5 | 271 | |||
XM_011533559.2 | XP_011531861.1 | 5, 6 | 440 | |||
XM_017006092.1 | XP_016861581.1 | 5, 6 | 332 | |||
XM_017006093.1 | XP_016861582.1 | 4, 5 | 322 | |||
XM_017006094.1 | XP_016861583.1 | 5, 6 | 355 | |||
GenBank mRNA | AF174589.1 | - | 5, 6 | 245 | ||
AF176518.1 | - | 4, 5 | 209 | |||
AF186273.1 | - | 4, 5 | 224 | |||
AK001271.1 | - | 4, 5 | 274 | |||
AK001438.1 | - | 4, 5 | 242 | |||
AK297534.1 | - | 4, 5 | 239 | |||
AK298967.1 | - | 4, 5 | 240 | |||
AK299665.1 | - | 5, 6 | 357 | |||
AK302218.1 | - | 6, 7 | 502 | |||
AK316383.1 | - | 5, 6 | 414 | |||
AU124816.1 | - | 4, 5 | 274 | |||
BC031556.1 | - | 4, 5 | 218 | 5197151 | ||
CR457121.1 | - | 4, 5 | 203 | |||
DA031508.1 | - | 4, 5 | 271 |