Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012836.2 | NP_036968.1 | 2 | 844 | ||
GenBank mRNA | AF284830.1 | - | 2 | 201 | ||
U62897.1 | - | 2 | 844 |