Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lipe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012859.1 | NP_036991.1 | 1 | 417 | ||
GenBank mRNA | BC078888.1 | - | 1 | 400 | ||
U40001.1 | - | 1 | 417 |