Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012872.1 | NP_037004.1 | 2, 3 | 153 | ||
GenBank mRNA | M33528.1 | - | 2, 3 | 153 | ||
M33530.1 | - | 2, 3 | 163 | |||
M60738.1 | - | 1, 2 | 126 |