Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012879.2 | NP_037011.2 | 11 | 1495 | ||
XM_006232207.2 | XP_006232269.1 | 12 | 1799 | |||
GenBank mRNA | BC078875.1 | - | 11 | 1495 | 7127751 | |
J03145.1 | - | 11 | 1570 |