Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012889.1 | NP_037021.1 | 654 | |||
GenBank mRNA | BC089812.1 | - | 672 | 7364929 | ||
M84488.1 | - | 654 | ||||
X63722.1 | - | 603 |
Gene Symbol: LOC100912479 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749384.3 | XP_003749432.1 | 3 | 867 |