Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083122.1 | NP_001076591.1 | 3 | 359 | ||
NM_012890.2 | NP_037022.1 | 2 | 298 | |||
GenBank mRNA | BC061997.1 | - | 2 | 301 | ||
BC168237.1 | - | 3 | 352 | 7938890 | ||
CB778290.1 | - | 3 | 362 | |||
U50927.1 | - | 2 | 298 |