Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012198.3 | NP_036330.1 | 5 | 503 | ||
XM_005246446.2 | XP_005246503.1 | 5 | 444 | |||
XM_006712398.3 | XP_006712461.1 | 5 | 676 | |||
XM_006712400.3 | XP_006712463.1 | 6 | 754 | |||
XM_006712401.3 | XP_006712464.1 | 5 | 755 | |||
XM_011510926.2 | XP_011509228.1 | 5 | 455 | |||
XM_011510927.2 | XP_011509229.1 | 5 | 450 | |||
XM_011510928.2 | XP_011509230.1 | 3 | 388 | |||
XM_017003764.1 | XP_016859253.1 | 6 | 767 | |||
XM_017003765.1 | XP_016859254.1 | 6 | 853 | |||
XM_017003766.1 | XP_016859255.1 | 6 | 650 | |||
XM_017003767.1 | XP_016859256.1 | 5 | 526 | |||
XM_017003768.1 | XP_016859257.1 | 5 | 521 | |||
GenBank mRNA | AB209768.1 | - | 5 | 392 | ||
AK295413.1 | - | 5 | 436 | |||
AK312349.1 | - | 5 | 452 | |||
BC005214.1 | - | 5 | 426 | 3996889 | ||
M81637.1 | - | 5 | 446 |