Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012910.2 | NP_037042.1 | 16 | 1282 | ||
XM_006229734.2 | XP_006229796.1 | 16 | 1908 | |||
XM_006229735.2 | XP_006229797.1 | 15 | 1883 | |||
XM_006229736.2 | XP_006229798.1 | 16 | 1877 | |||
XM_006229737.2 | XP_006229799.1 | 15 | 1852 | |||
XM_006229739.2 | XP_006229801.1 | 16 | 2024 | |||
XM_006229740.2 | XP_006229802.1 | 16 | 1410 | |||
XM_006229741.2 | XP_006229803.1 | 16 | 1423 | |||
GenBank mRNA | M91589.1 | - | 16 | 1285 |