Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012203.1 | NP_036335.1 | 6 | 619 | ||
XM_005251631.1 | XP_005251688.1 | 3 | 312 | |||
XM_011518073.2 | XP_011516375.1 | 6 | 993 | |||
XM_017015320.1 | XP_016870809.1 | 6 | 663 | |||
XM_017015321.1 | XP_016870810.1 | 6 | 663 | |||
XM_017015322.1 | XP_016870811.1 | 3 | 312 | |||
XM_017015323.1 | XP_016870812.1 | 6 | 993 | |||
GenBank mRNA | AF134895.1 | - | 6 | 619 | ||
AF146018.1 | - | 6 | 618 |