Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012928.1 | NP_037060.1 | 2282 | |||
GenBank mRNA | AF126808.1 | - | 2176 | |||
X55519.1 | - | 2282 |
Gene Symbol: LOC103690077 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008759081.1 | XP_008757303.1 | 3 | 1634 |