Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012932.2 | NP_037064.1 | 3 | 363 | ||
XM_006251084.2 | XP_006251146.1 | 3 | 597 | |||
GenBank mRNA | FM031363.1 | - | 3 | 363 | ||
U52095.1 | - | 3 | 157 | |||
U52102.1 | - | 3 | 331 |