Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crybb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012937.1 | NP_037069.1 | 4 | 352 | ||
GenBank mRNA | X16072.1 | - | 4 | 352 |