Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sparcl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012946.1 | NP_037078.1 | 3 | 503 | ||
GenBank mRNA | BC061755.1 | - | 3 | 504 | 5600008 | |
U27562.1 | - | 3 | 503 |