Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eno3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012949.2 | NP_037081.2 | 5, 6 | 358 | ||
XM_006246599.2 | XP_006246661.1 | 5, 6 | 429 | |||
XM_006246600.2 | XP_006246662.1 | 5, 6 | 409 | |||
GenBank mRNA | BC083566.1 | - | 5, 6 | 345 | 7193009 | |
DY321069.1 | - | 5, 6 | 358 | |||
FQ214810.1 | - | 4, 5 | 336 | |||
FQ214945.1 | - | 5, 6 | 376 | |||
FQ214984.1 | - | 5, 6 | 368 | |||
FQ215034.1 | - | 1, 2 | 35 | |||
FQ215197.1 | - | 1, 2 | 35 | |||
FQ215221.1 | - | 4, 5 | 271 | |||
FQ215310.1 | - | 1, 2 | 35 | |||
FQ215357.1 | - | 5, 6 | 380 | |||
FQ215526.1 | - | 5, 6 | 368 | |||
FQ215607.1 | - | 1, 2 | 35 | |||
FQ215648.1 | - | 5, 6 | 368 | |||
FQ215780.1 | - | 1, 2 | 35 | |||
FQ215843.1 | - | 4, 5 | 335 | |||
FQ215865.1 | - | 5, 6 | 368 | |||
FQ216020.1 | - | 1, 2 | 35 | |||
FQ216150.1 | - | 4, 5 | 336 | |||
FQ216271.1 | - | 1, 2 | 35 | |||
FQ216382.1 | - | 5, 6 | 368 | |||
FQ216386.1 | - | 4, 5 | 345 | |||
FQ216660.1 | - | 4, 5 | 335 | |||
FQ216781.1 | - | 1, 2 | 35 | |||
FQ217095.1 | - | 5, 6 | 368 | |||
FQ223831.1 | - | 1, 2 | 35 | |||
FQ224938.1 | - | 5, 6 | 368 | |||
Y00979.1 | - | 4, 5 | 320 |