Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012952.1 | NP_037084.1 | 3 | 831 | ||
GenBank mRNA | D14839.1 | - | 3 | 850 |