Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012959.1 | NP_037091.1 | 2 | 395 | ||
XM_008760512.1 | XP_008758734.1 | 3 | 538 | |||
XM_008760513.1 | XP_008758735.1 | 3 | 402 | |||
XM_008760514.1 | XP_008758736.1 | 2 | 433 | |||
GenBank mRNA | AJ002072.1 | - | 2 | 139 | ||
U59486.1 | - | 2 | 440 | |||
U97142.1 | - | 2 | 395 |
Gene Symbol: LOC100911751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231658.2 | XP_006231720.1 | 3 | 424 | ||
XM_006231659.2 | XP_006231721.1 | 3 | 431 | |||
XM_006231660.2 | XP_006231722.1 | 3 | 542 | |||
XM_006231661.2 | XP_006231723.1 | 3 | 508 | |||
XM_006231662.2 | XP_006231724.1 | 3 | 406 | |||
XM_008760583.1 | XP_008758805.1 | 3 | 409 |