Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012959.1 | NP_037091.1 | 2, 3 | 597 | ||
XM_008760512.1 | XP_008758734.1 | 3, 4 | 740 | |||
XM_008760513.1 | XP_008758735.1 | 3, 4 | 604 | |||
XM_008760514.1 | XP_008758736.1 | 2, 3 | 635 | |||
GenBank mRNA | AJ002072.1 | - | 2, 3 | 341 | ||
U59486.1 | - | 2, 3 | 642 | |||
U97142.1 | - | 2, 3 | 597 |
Gene Symbol: LOC100911751 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231658.2 | XP_006231720.1 | 3, 4 | 626 | ||
XM_006231659.2 | XP_006231721.1 | 3, 4 | 633 | |||
XM_006231660.2 | XP_006231722.1 | 3, 4 | 744 | |||
XM_006231661.2 | XP_006231723.1 | 3, 4 | 710 | |||
XM_006231662.2 | XP_006231724.1 | 3, 4 | 608 | |||
XM_008760583.1 | XP_008758805.1 | 3, 4 | 611 |