Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012227.3 | NP_036359.3 | 2 | 506 | ||
XM_006724868.3 | XP_006724931.1 | 3 | 406 | |||
GenBank mRNA | AK024630.1 | - | 2 | 445 | ||
AK223412.1 | - | 2 | 451 | |||
AK296003.1 | - | 2 | 167 | |||
AK298260.1 | - | 2 | 253 | |||
BC014636.1 | - | 2 | 179 | 3528598 | ||
BG437484.1 | - | 2 | 589 | |||
BT007360.1 | - | 1 | 135 | |||
CN356690.1 | - | 2 | 471 | |||
DN997384.1 | - | 2 | 426 | |||
Y14391.2 | - | 2 | 464 |