Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012980.2 | NP_037112.2 | 2 | 195 | ||
GenBank mRNA | BC099781.1 | - | 2 | 189 | 7933545 | |
U46034.1 | - | 2 | 195 |
Gene Symbol: LOC103694874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773828.1 | XP_008772050.1 | 2 | 221 |