Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012988.2 | NP_037120.1 | 9 | 1647 | ||
GenBank mRNA | AB060652.1 | - | 1555 | |||
AF112455.1 | - | 1727 | ||||
D78017.1 | - | 9 | 1647 | |||
D78018.1 | - | 8 | 1576 | |||
D78019.1 | - | 5 | 1240 | |||
X13167.1 | - | 9 | 1489 |