Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013012.1 | NP_037144.1 | 4 | 754 | ||
GenBank mRNA | Z36276.1 | - | 4 | 754 |