Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pspn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013014.1 | NP_037146.1 | 2 | 388 | ||
GenBank mRNA | AF040961.1 | - | 2 | 388 |