Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013024.2 | NP_037156.2 | 4 | 584 | ||
XM_008767547.1 | XP_008765769.1 | 3 | 570 | |||
XM_008767548.1 | XP_008765770.1 | 3 | 390 | |||
GenBank mRNA | CA339008.1 | - | 4 | 584 | ||
J02675.1 | - | 3 | 594 |