Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc18a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013031.1 | NP_037163.1 | 16 | 1767 | ||
XM_006231664.2 | XP_006231726.1 | 16 | 2608 | |||
XM_008760515.1 | XP_008758737.1 | 14 | 2099 | |||
XM_008760516.1 | XP_008758738.1 | 15 | 2745 | |||
XM_008760517.1 | XP_008758739.1 | 15 | 2740 | |||
XM_008760519.1 | XP_008758741.1 | 13 | 2006 | |||
XM_008760520.1 | XP_008758742.1 | 12 | 1922 | |||
GenBank mRNA | L00603.1 | - | 16 | 1767 |