Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scgb1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013051.1 | NP_037183.1 | 3 | 338 | ||
GenBank mRNA | BC069174.1 | - | 3 | 328 | 7096714 | |
J05536.1 | - | 3 | 338 |