Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhaq | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013053.1 | NP_037185.1 | 6 | 1106 | ||
GenBank mRNA | BC062409.1 | - | 5 | 1076 | 5623850 | |
D17614.1 | - | 6 | 1106 | |||
FQ218679.1 | - | 5 | 866 | |||
FQ219995.1 | - | 2 | 393 | |||
FQ232769.1 | - | 1 | 173 |