Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Id3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013058.2 | NP_037190.1 | 3 | 665 | ||
GenBank mRNA | BC064658.1 | - | 3 | 665 | 5623701 | |
FQ212997.1 | - | 3 | 687 | |||
FQ216267.1 | - | 3 | 687 | |||
FQ218468.1 | - | 3 | 685 | |||
FQ218958.1 | - | 3 | 678 | |||
FQ229332.1 | - | 3 | 687 |