Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNPEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319116.1 | NP_001306045.1 | 8 | 858 | ||
NM_001319118.1 | NP_001306047.1 | 8 | 786 | |||
NM_001319119.1 | NP_001306048.1 | 8 | 1066 | |||
NM_001319121.1 | NP_001306050.1 | 9 | 759 | |||
NM_001319122.1 | NP_001306051.1 | 8 | 851 | |||
NM_012100.3 | NP_036232.2 | 8 | 883 | |||
GenBank mRNA | AF005050.2 | - | 8 | 858 | ||
AK001777.1 | - | 8 | 883 | |||
AK001977.1 | - | 8 | 855 | |||
AK021871.1 | - | 8 | 735 | |||
AK023005.1 | - | 8 | 797 | |||
AK301919.1 | - | 8 | 843 | |||
AK302809.1 | - | 8 | 850 | |||
AL519865.3 | - | 8 | 829 | |||
BC000653.2 | - | 8 | 765 | 3343505 | ||
BC004854.2 | - | 4 | 416 | |||
BX403948.2 | - | 8 | 914 |