Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013073.3 | NP_037205.2 | 2 | 222 | ||
GenBank mRNA | BC088417.1 | - | 2 | 222 | 7123213 | |
D11475.1 | - | 2 | 82 |