Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptprz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170685.1 | NP_001164156.1 | 4 | 470 | ||
NM_013080.2 | NP_037212.2 | 4 | 470 | |||
XM_006236137.2 | XP_006236199.1 | 4 | 509 | |||
XM_006236138.2 | XP_006236200.1 | 4 | 509 | |||
XM_006236139.2 | XP_006236201.1 | 4 | 509 | |||
GenBank mRNA | U04998.1 | - | 4 | 470 | ||
U09357.1 | - | 4 | 470 |