Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadsb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013084.2 | NP_037216.1 | 4, 5 | 520 | ||
XM_008759865.1 | XP_008758087.1 | 4, 5 | 605 | |||
XM_008759866.1 | XP_008758088.1 | 4, 5 | 605 | |||
XM_008759867.1 | XP_008758089.1 | 4, 5 | 606 | |||
GenBank mRNA | FQ210776.1 | - | 4, 5 | 529 | ||
U64451.1 | - | 4, 5 | 521 |