Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AHSA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321441.1 | NP_001308370.1 | 9 | 1221 | ||
NM_012111.2 | NP_036243.1 | 9 | 1355 | |||
GenBank mRNA | AF161440.1 | - | 8 | 999 | ||
AF164791.1 | - | 9 | 1259 | |||
AJ243310.1 | - | 9 | 1326 | |||
BC000321.2 | - | 9 | 1303 | 2821759 | ||
BC007398.2 | - | 8 | 967 |