Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012112.4 | NP_036244.2 | 12 | 2059 | ||
XM_011528697.2 | XP_011526999.1 | 12 | 1617 | |||
XM_011528699.2 | XP_011527001.1 | 11 | 1805 | |||
XM_011528700.2 | XP_011527002.1 | 11 | 1538 | |||
GenBank mRNA | AB024704.1 | - | 12 | 1832 | ||
AB027467.1 | - | 11 | 1468 | |||
AF098158.1 | - | 12 | 1852 | |||
AF146731.1 | - | 11 | 1465 | |||
AF244547.1 | - | 9 | 1239 | |||
AF287265.1 | - | 13 | 1933 | |||
AK001391.1 | - | 8 | 1142 | |||
AK312549.1 | - | 12 | 1837 | |||
AL117534.1 | - | 10 | 1303 | |||
AY734680.1 | - | 10 | 1361 | |||
BC004136.1 | - | 12 | 1789 | 3509275 | ||
BC020207.1 | - | 12 | 1818 | 5015279 |