Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013101.3 | NP_037233.3 | 8 | 1048 | ||
GenBank mRNA | AF110461.2 | - | 8 | 740 | ||
L27057.1 | - | 8 | 1048 | |||
L27062.1 | - | 2 | 578 | |||
L36467.1 | - | 938 | ||||
M26715.1 | - | 2 | 312 | |||
M26716.1 | - | 2 | 216 | |||
M26717.1 | - | 2 | 280 |