Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013102.3 | NP_037234.2 | 5 | 727 | ||
GenBank mRNA | BC070519.1 | - | 5 | 730 | 7103884 | |
BC126071.1 | - | 5 | 685 | 8363608 | ||
FQ214119.1 | - | 6 | 713 | |||
FQ228836.1 | - | 4 | 703 |