Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013106.1 | NP_037238.1 | 5, 6 | 671 | ||
GenBank mRNA | DQ120473.1 | - | 5, 6 | 564 | ||
DQ120474.1 | - | 5, 6 | 564 | |||
FQ222046.1 | - | 5, 6 | 734 | |||
J03219.1 | - | 5, 6 | 680 | |||
M20713.1 | - | 5, 6 | 671 |