Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013111.3 | NP_037243.2 | 692 | |||
XM_008768943.1 | XP_008767165.1 | 2 | 964 | |||
XM_008768944.1 | XP_008767166.1 | 2 | 964 | |||
GenBank mRNA | AB066224.1 | - | 2 | 449 | ||
AF245000.1 | - | 632 | ||||
D67087.1 | - | 2 | 422 | |||
L10152.1 | - | 2 | 416 | |||
U70476.1 | - | 2 | 501 |