Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apoa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013112.1 | NP_037244.1 | 3 | 250 | ||
XM_006250238.2 | XP_006250300.1 | 4 | 384 | |||
XM_006250239.2 | XP_006250301.1 | 4 | 390 | |||
GenBank mRNA | FQ209753.1 | - | 4 | 315 | ||
FQ209821.1 | - | 4 | 314 | |||
FQ210214.1 | - | 4 | 308 | |||
FQ210245.1 | - | 4 | 315 | |||
FQ210295.1 | - | 4 | 312 | |||
FQ210325.1 | - | 4 | 335 | |||
FQ210462.1 | - | 4 | 313 | |||
FQ210537.1 | - | 3 | 309 | |||
FQ210542.1 | - | 3 | 438 | |||
FQ210648.1 | - | 4 | 312 | |||
FQ210885.1 | - | 4 | 342 | |||
FQ210979.1 | - | 4 | 315 | |||
FQ211006.1 | - | 4 | 313 | |||
FQ211265.1 | - | 4 | 312 | |||
FQ211383.1 | - | 4 | 314 | |||
FQ218273.1 | - | 4 | 315 | |||
FQ218326.1 | - | 4 | 309 | |||
FQ218756.1 | - | 4 | 316 | |||
FQ221741.1 | - | 4 | 315 | |||
M28615.1 | - | 3 | 219 | |||
X03468.1 | - | 3 | 250 |