Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anxa5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013132.1 | NP_037264.1 | 13 | 1376 | ||
XM_006232268.2 | XP_006232330.1 | 13 | 1513 | |||
GenBank mRNA | BC081855.1 | - | 13 | 1431 | 7192977 | |
FQ213209.1 | - | 14 | 1508 | |||
FQ214176.1 | - | 13 | 1410 | |||
FQ229127.1 | - | 13 | 1366 | |||
M21730.1 | - | 13 | 1376 |