Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgcr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013134.2 | NP_037266.2 | 6 | 519 | ||
XM_006231826.2 | XP_006231888.1 | 6 | 927 | |||
GenBank mRNA | BC064654.1 | - | 6 | 519 | 5600023 |